Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BSA Monoclonal Antibody (A7D10), Huabio
Mouse Monoclonal Antibody
Supplier: HUABIO RESEARCH INC HA600085


Description
BSA Monoclonal antibody specifically detects BSA in Chemical samples. It is validated for ELISA, Western BlotSpecifications
BSA | |
Monoclonal | |
2 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
A7D10 | |
Unconjugated | |
ALB; ALB protein; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; Bos d 6; bovine albumin; bovine serum albumin; BSA; cell growth inhibiting protein 42; FDAH; GIG20; GIG42; growth-inhibiting protein 20; MGC179277; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
Native protein. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction