Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BSA Mouse, anti-Bovine, HRP, Clone: 2D11G3, Proteintech
SDP

Catalog No. 502258357 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-8357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-8357 Supplier Proteintech Group Inc Supplier No. HRP66201100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BSA, also named as ALB and Bovine Serum albumin, is the main protein of plasma. It has a good binding capacity for water, Ca2+, Na+, K+, fatty acids, hormones, bilirubin and drugs. Its main function is the regulation of the colloidal osmotic pressure of blood.

Specifications

Antigen BSA
Applications ELISA
Classification Monoclonal
Clone 2D11G3
Concentration 1000 μg/mL
Conjugate HRP
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Target Species Bovine
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.