Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BSAP/PAX5 Mouse anti-Human, Clone: 2H2D6, Proteintech
SDP

Catalog No. 501736436 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6436 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6436 Supplier Proteintech Group Inc Supplier No. 603491IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is specific to PAX5. It will not cross react with other PAX members.

PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.

Specifications

Antigen BSAP/PAX5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2H2D6
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PAX5
Gene Accession No. Q02548
Gene Alias BSAP, BSAP,PAX5, paired box 5, Paired box protein Pax 5, PAX5
Gene Symbols PAX5
Host Species Mouse
Immunogen BSAP,PAX5 Fusion Protein Ag16023
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5079
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.