Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BSDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556712 Supplier Invitrogen™ Supplier No. PA556712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLRVFELNSD SGKSTPSNNG KKGSSTDISE DWEKDFDLDM TEEEVQMALS KVDASGEVSG PGGSEGSEPN GPGCESSPQP AQLSPQEGPC SCL Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 60%.

BSDC1 (BSD Domain Containing 1) is a Protein Coding gene. An important paralog of this gene is SYAP1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen BSDC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BSDC1
Gene Accession No. Q9NW68
Gene Alias 1110063F24Rik; AW011758; BSD domain containing 1; BSD domain-containing protein 1; BSDC1; DKFZp686B09139; FLJ10276; hypothetical protein LOC505545; RGD1311622; RP4-811H24.7; UNQ2494/PRO5781
Gene Symbols BSDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BSDC1. Recombinant protein control fragment (Product #RP-102592).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55108
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.