Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BSND Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596786 Supplier Invitrogen™ Supplier No. PA596786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CQCYPKITFV PADSDFQGIL SPKAMGLLEN GLAAEMKSPS PQPPYVRLWE EAAYDQSLPD FSHIQMKVMS YSEDHRSLLA PEMGQPKLGT SDGGEGGPGD VQAWMEAAVV IHKGSDESEG ERRLTQSWPG PLACPQGPAP LASFQDDLDM DSSEGSSPNA SPHDREEACS PQQEPQGCRC PLDRFQDFAL IDAPTLEDEP QEGQQWEIAL PNNWQRYPRT KVEEKEASDT GGEEPEKEEE DLYYGLPDGA GDLLPDKELG FEPDTQG; Positive Samples: 293T, Rat liver, Rat kidney, Rat small intestine; Cellular Location: Cell membrane, Cytoplasm, Multi-pass membrane protein.

BSND encodes an essential beta subunit for CLC chloride channels. These heteromeric channels localize to basolateral membranes of renal tubules and of potassium-secreting epithelia of the inner ear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BSND
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BSND
Gene Accession No. Q8R2H3, Q8VIM4, Q8WZ55
Gene Alias BART; Bartter syndrome, infantile, with sensorineural deafness (Barttin); barttin; barttin CLCNK type accessory beta subunit; barttin CLCNK-type chloride channel accessory beta subunit; Bsnd; deafness, autosomal recessive 73; DFNB73
Gene Symbols BSND
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 54-320 of human BSND (NP_4765171).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140475, 192675, 7809
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.