Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BSPH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583908 Supplier Invitrogen™ Supplier No. PA583908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SACIFPVILN ELSSTVETIT HFPEVTDGEC VFPFHYKNGT YYDCIKSKAR HKWCSLNKTY Highest antigen sequence indentity to the following orthologs: Rat - 43%, Mouse - 42%.

BSPH1 is a protein coding gene. Binds sperm in vitro and promotes sperm capacitation. Specifically promotes capacitation induced by high density lipoproteins (HDLs). Also binds heparin, phospholipid liposomes, and weakly to gelatin. Does not bind chondroitin sulfate B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BSPH1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BSPH1
Gene Accession No. Q075Z2
Gene Alias binder of sperm 1; binder of sperm protein homolog 1; Bovine seminal plasma protein homolog 1; bovine seminal plasma protein-like 1; BSP1; BSPH1; ELSPBP2; epididymal sperm binding protein 2
Gene Symbols BSPH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BSPH1. Recombinant protein control fragment (Product #RP-99199).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131137
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.