Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BST-2 (CD317), Rat anti-Mouse, Clone: 927, Millipore Sigma™

Catalog No. MABF204725 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF204725 25 μg
MABF2047100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF204725 Supplier MilliporeSigma Supplier No. MABF204725UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-BST-2 (CD317), clone 927, Cat. No. MABF2047, is a rat monoclonal antibody that detects Bone marrow stromal antigen 2 and has been tested for use in ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), and Neutralizing activity.

Bone marrow stromal antigen 2 (UniProt: Q8R2Q8; also known as BST-2, HM1.24 antigen, PDCA-1, CD317) is encoded by the Bst2 gene (Gene ID: 69550) in murine species. BST-2 is a single-pass type II disulfide-linked homodimeric membrane protein that shuttles between the cell membrane, where it is present predominantly in membrane/lipid rafts, and the trans-Golgi network. Its dimerization is essential for its antiviral activity. BST-2 is also shown to form a complex with MMP-14 and localize to the cytoplasm. BST-2 is highly expressed on type I interferon-producing cells in naïve mice. Its levels in most cells are elevated following treatment with type I IFNs and IFN-gamma. It serves as an IFN-induced antiviral host restriction factor that efficiently blocks the release of diverse mammalian enveloped viruses by directly tethering nascent virions to the membranes of infected cells. It acts as a direct physical tether, holding virions to the cell membrane and linking virions to each other. The tethered virions can be either internalized by endocytosis and subsequently degraded or they can remain on the cell surface. In either case, their spread as cell-free virions is restricted. BST-2 is synthesized with a propeptide (aa 153-172), which is removed to generate the mature form. The mature form has a cytoplasmic domain (aa 1-30), a transmembrane domain (aa 31-51), and the extracellular domain (aa 52-152). Within the extracellular region, it has a coiled coil domain (aa 74-147) that forms an extended 17 nm long semi-flexible rod-like structure, which is essential for virion retention at the cell surface and prevent virus spreading.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BST-2 (CD317)
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Neutralization
Classification Monoclonal
Clone 927
Conjugate Unconjugated
Dilution Neutralizing Analysis: A representative lot efficiently depleted type I interferon-producing cells (IPC) and abrogated IFN-alpha secretion by IPC in response to CpG. (Blasius, A.L., et. al. (2006). J Immunol. 177(5):3260-5).Immunohistochemistry (Paraffin) Analysis: A representative lot detected BST-2 (CD317) in Immunohistochemistry applications (Yun, T., et. al. (2016). Cell Metab. 23(5):852-66).ELISA Analysis: A representative lot detected BST-2 (CD317) in ELISA applications (Blasius, A.L., et. al. (2006). J Immunol. 177(5):3260-5).Flow Cytometry Analysis: A representative lot detected BST-2 (CD317) in Flow Cytometry applications (Blasius, A.L., et. al. (2006). J Immunol. 177(5):3260-5; Yun, T., et. al. (2016). Cell Metab. 23(5):852-66).
Formulation Purified rat monoclonal antibody IgG2b in PBS without azide.
Gene Alias Bone marrow stromal antigen 2;BST-2;HM1.24 antigen;CD317
Gene Symbols Bst2
Host Species Rat
Immunogen Type I IFN-producing cells derived from mouse bone marrow.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_932763.1
Test Specificity Clone 927 is a rat monoclonal antibody that specifically detects murine Bone marrow stromal antigen 2 (BST-2). It targets an epitope with in the extracellular domain.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.