Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BST2 Antibody (2E2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $426.00
Specifications
| Antigen | BST2 |
|---|---|
| Clone | 200 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 ug/ml, Flow Cytometry 0.5 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP229621SS
![]() |
Novus Biologicals
NBP229621SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP229621
![]() |
Novus Biologicals
NBP229621 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
BST2 Monoclonal specifically detects BST2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| BST2 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| Q10589 | |
| 684 | |
| Full length recombinant human BST2 protein [NCBI NP_004326] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 200 | |
| Western Blot 1 ug/ml, Flow Cytometry 0.5 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST-2, CD317, CD317 antigen, HM1.24 antigen, NPC-A-7, PDCA-1, TETHERIN | |
| BST2 | |
| IgG1 κ | |
| Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title