Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BST2 Antibody (4F6), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP229622APCCY7
Description
BST2 Monoclonal specifically detects BST2 in Human samples. It is validated for Flow Cytometry.Specifications
BST2 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
BST2 | |
Full length recombinant human BST2 protein [NCBI NP_004326]. | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
4F6 | |
Flow Cytometry | |
bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST-2, CD317, CD317 antigen, HM1.24 antigen, NPC-A-7, PDCA-1, TETHERIN | |
Mouse | |
Protein A or G purified | |
Signal Transduction | |
684.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction