Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse BST-2/Tetherin Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. FAB8660S100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8660S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8660S100 Supplier R&D Systems Supplier No. FAB8660S100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Antibody

BST2 Monoclonal specifically detects BST2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen BST2
Applications Flow Cytometry
Classification Monoclonal
Clone 44E9R
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST2, BST-2, CD317, CD317 antigen, HM1.24 Antigen, NPC-A-7, PDCA-1, Tetherin
Host Species Rat
Immunogen Recombinant extracellular domain of CD317
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 684
Test Specificity Detects mouse BST-2/Tetherin in flow cytometry.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.