Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent

Catalog No. PI38840
Encompass
Change view
Click to view available options
Standard Components:
BSTFA +1% TMCS
BSTFA +10% TMCS
Quantity:
10 g
10 x 1 mL Ampules
100 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Components Quantity
PI38840 BSTFA +10% TMCS 10 x 1 mL Ampules
PI38831 BSTFA +1% TMCS 10 x 1 mL Ampules
PI38832 BSTFA +1% TMCS 10 g
PI38833 BSTFA +1% TMCS 25 g
PI38834 BSTFA +1% TMCS 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PI38840 Supplier Thermo Scientific™ Supplier No. TS38840
Add to Cart
Add to Cart

Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent is derivatizing compounds.

Use Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent for analytes that are difficult to silylate. Stronger than BSTFA alone, this catalyzed formulation is ideal for derivatizing compounds such as fatty acid amides and slightly hindered hydroxyls for GC analysis.

  • Excellent for derivatizing fatty acid amides, slightly hindered hydroxyls and other compounds
  • Catalyzed formulation is stronger than BSTFA alone
TRUSTED_SUSTAINABILITY

Specifications

Item Description BSTFA +10% TMCS; 10 x 1 mL Ampules
Storage 2°C to 8°C
Quantity 10 x 1 mL Ampules
Standard Components BSTFA +10% TMCS

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.