Learn More
Bsu DNA Polymerase Large Fragment, Glycerol Free, G-Biosciences
Bsu DNA Polymerase, large fragment is from Bacillus subtilis.
Supplier: G-Biosciences 7862149
Description
It is modified to retain its 5'-to-3' DNA polymerase activity with its 5'-to-3' exonuclease domain removed. It also lacks the 3'-to-5' exonuclease activity.
Specifications
Gel Electrophoresis | |
High | |
20 min. | |
DNA Polymerase | |
Microbial Detection, Pathogen Detection, Isothermal Amplification |
Liquid | |
End-point PCR | |
Biotin, FAM, TAMRA | |
200 U |