Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BSX Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103664
Description
Antibody detects endogenous levels of total BSX.
BSX is a DNA binding protein that functions as transcriptional activator. It is essentiel for normal postnatal growth and nursing. It is also an essential factor for neuronal neuropeptide Y and agouti-related peptide function and locomotory behavior in the control of energy balance.
Specifications
BSX | |
Polyclonal | |
Unconjugated | |
BSX | |
brain specific homeobox; brain-specific homeobox protein homolog; Bsx; BSX1; Bsx1a; Bsx1b; RGD1565120 | |
Rabbit | |
Affinity chromatography | |
RUO | |
244813, 390259, 500982 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q3C1V8, Q810B3 | |
BSX | |
A synthesized peptide derived from human BSX(Accession Q3C1V8), corresponding to amino acid residues E193-L233. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction