Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTAF1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10057501R
Description
BTAF1 Polyclonal antibody specifically detects BTAF1 in Human samples. It is validated for Western BlotSpecifications
BTAF1 | |
Polyclonal | |
Western Blot | |
ATP-dependent helicase BTAF1, BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1homolog, S. cerevisiae), B-TFIID transcription factor-associated 170 kDa subunit, EC 3.6.1, EC 3.6.4.-, MOT1MGC138406, TAF(II)170KIAA0940, TAF-172, TAF172BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170 kD (Mot1homolog, S. cerevisiae), TAFII170TATA-binding protein-associated factor 172, TBP-associated factor 172 | |
The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human RNA polymerase II, B-TFIID transcription factor-associated, 170kDa using the numbering given in entry NP_003963.1 (GeneID 9044). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9044 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction