Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTAF1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501559156
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9156 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9156 Supplier Bethyl Laboratories, Inc Supplier No. A301065AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Initiation of transcription by RNA polymerase II requires the assistance of TATA box-binding protein (TBP; MIM 600075) and TBP-associated factors, or TAFs (e.g., TAF2B; MIM 604912), in 2 distinct complexes, TFIID and B-TFIID. The TFIID complex is composed of TBP and more than 8 TAFs. However, the majority of TBP is present in the B-TFIID complex, which is composed of TBP and TAFII170, also called TAF172, and has DNA-dependent ATPase activity.

Specifications

Antigen BTAF1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene BTAF1
Gene Accession No. O14981
Gene Alias MOT1, TAF(II)170, TAF172, TAFII170, ATP-dependent helicase BTAF1, B-TFIID transcription factor-associated 170 kDa subunit, TATA-binding protein-associated factor 172, TBP-associated factor 172
Gene Symbols BTAF1
Host Species Rabbit
Immunogen Between 1325 and 1375
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9044
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.