Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ BTB1
GREENER_CHOICESDP

Catalog No. 539950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-995-0 50 mg
53-991-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-995-0 Supplier R&D Systems™ Supplier No. 5399/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective and ATP-competitive Kif18A inhibitor

BTB1 is a selective and ATP-competitive mitotic kinesin Kif18A inhibitor (IC50 = 1.7 μM); reversibly inhibits microtubule-stimulated Kif18A ATPase activity. Displays selectivity against a panel of other mitotic kinesins.

Specifications

Quantity 50 mg
Product Line Tocris
Product Type BTB1
Purity >99%
Molecular Weight (g/mol) 297.71
Inhibitors BTB1

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.