Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTBD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31769025UL
This item is not returnable.
View return policy
Description
BTBD1 Polyclonal antibody specifically detects BTBD1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| BTBD1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| BTB (POZ) domain containing 1, BTB domain containing 1, BTB/POZ domain-containing protein 1, C15orf1, HCV NS5A-transactivated protein 8, Hepatitis C virus NS5A-transactivated protein 8, NS5ATP8 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: PSPSSLGPLLPLQREPLYNWQATKASLKERFAFLFNSELLSDVRFVLGKG | |
| 25 μg | |
| Cell Biology, Cell Cycle and Replication | |
| 53339 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction