Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTBD10 Monoclonal Antibody (OTI7D3)
GREENER_CHOICE

Catalog No. PIMA526985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526985 Supplier Invitrogen™ Supplier No. MA526985
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BTBD10 Monoclonal Antibody for Western Blot, IHC (P)

BTBD10 appears to behave as a suppressor of cell death, which includes neuronal cell death related to amyotrophic lateral sclerosis. It may also act as an enhancer of cell growth via its positive regulation of Akt phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BTBD10
Gene Accession No. Q9BSF8
Gene Alias 1110056N09Rik; BTB (broad-complex tramtrack and bric-a-brac/pox virus and zinc finger) domain-containing protein 10; BTB (POZ) domain containing 10; BTB domain containing 10; BTB/POZ domain-containing protein 10; Btbd10; Glucose metabolism-related protein 1; Gmrp1; GMRP-1; K+ channel tetramerization protein; RGD1306301
Gene Symbols BTBD10
Host Species Mouse
Immunogen Full length human recombinant protein of BTBD10 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84280
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.