Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTBD10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561320
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561320 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561320 Supplier Invitrogen™ Supplier No. PA561320
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKLHSRPRKL YKHSSTSSRI AKGGVDHTKM SLHGASGGHE RSRDRRRSSD RSRDSSHERT ESQLTPCIRN VTSPTRQHHV EREKDHSSSR PS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 79%.

BTBD10 appears to behave as a suppressor of cell death, which includes neuronal cell death related to amyotrophic lateral sclerosis. It may also act as an enhancer of cell growth via its positive regulation of Akt phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BTBD10
Gene Accession No. Q9BSF8
Gene Alias 1110056N09Rik; BTB (broad-complex tramtrack and bric-a-brac/pox virus and zinc finger) domain-containing protein 10; BTB (POZ) domain containing 10; BTB domain containing 10; BTB/POZ domain-containing protein 10; Btbd10; Glucose metabolism-related protein 1; Gmrp1; GMRP-1; K+ channel tetramerization protein; RGD1306301
Gene Symbols BTBD10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BTBD10. Recombinant protein control fragment (Product #RP-102998).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84280
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.