Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTBD10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988529
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8529 Supplier Bioss Supplier No. BS8398R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTBD10 appears to behave as a suppressor of cell death, which includes neuronal cell death related to amyotrophic lateral sclerosis. It may also act as an enhancer of cell growth via its positive regulation of Akt phosphorylation.

Specifications

Antigen BTBD10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BTBD10
Gene Accession No. Q9BSF8
Gene Alias 1110056N09Rik; BTB (broad-complex tramtrack and bric-a-brac/pox virus and zinc finger) domain-containing protein 10; BTB (POZ) domain containing 10; BTB domain containing 10; BTB/POZ domain-containing protein 10; Btbd10; Glucose metabolism-related protein 1; GMRP1; GMRP-1; K+ channel tetramerization protein
Gene Symbols BTBD10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BTBD10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84280
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.