Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTBD11 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA565987
Description
Immunogen sequence: RRVPVLGSLLN LPGIRSFVDK VGESN Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Specifications
BTBD11 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6QL63 | |
BTBD11 | |
Recombit Human BTBD11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
BTBD11 | |
ABTB2B; ankyrin repeat and BTB/POZ domain-containing protein BTBD11; ANK-repeat BTB domain containing protein; BTB (POZ) domain containing 11; BTB/POZ domain-containing protein 11 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
121551 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction