Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTBD12 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128679AF700
Description
BTBD12 Polyclonal antibody specifically detects BTBD12 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
BTBD12 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
BTB (POZ) domain containing 12, BTB/POZ domain-containing protein 12, BTBD12structure-specific endonuclease subunit SLX4, KIAA1784MUS312, KIAA1987FANCP, SLX4 structure-specific endonuclease subunit homolog (S. cerevisiae) | |
The immunogen recognized by this antibody maps to a region between residue 1650 and 1700 of human BTB (POZ) domain containing 12 using the numbering given in entry NP_115820.2 (GeneID 84464). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
84464 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction