Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTBD12 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128680AF750
Description
BTBD12 Polyclonal specifically detects BTBD12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
BTBD12 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
BTB (POZ) domain containing 12, BTB/POZ domain-containing protein 12, BTBD12structure-specific endonuclease subunit SLX4, KIAA1784MUS312, KIAA1987FANCP, SLX4 structure-specific endonuclease subunit homolog (S. cerevisiae) | |
Rabbit | |
Affinity Purified | |
Primary | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
SLX4 | |
The immunogen recognized by this antibody maps to a region between residue 1784 to 1834 of human BTB (POZ) domain containing 12 using the numbering given in entry NP_115820.2 (GeneID 84464). | |
0.1 mL | |
84464.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction