Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTBD12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | BTBD12 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
BTBD12 Polyclonal specifically detects BTBD12 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
BTBD12 | |
Polyclonal | |
Rabbit | |
Human | |
84464 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LIQYVNNEGFSAVEDGVLTQRVLLGDVSTEAARTFLHYLYTADTGLPPGLSSELSSLAHRFGVSELVHLCEQVPIATDSEGKPWEEKEAENCESRA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
BTB (POZ) domain containing 12, BTB/POZ domain-containing protein 12, BTBD12structure-specific endonuclease subunit SLX4, KIAA1784MUS312, KIAA1987FANCP, SLX4 structure-specific endonuclease subunit homolog (S. cerevisiae) | |
SLX4 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title