Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BTBD12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545039
Description
Peptide sequence: TKTLQGPAEK KPPSGSQAPR TKKQRVTKWQ ASEPAHSVNG EGGVLASAPD Sequence homology: Human: 100%; Rabbit: 91%.
This gene encodes a structure-specific endonuclease subunit. The encoded protein contains a central BTB domain and it forms a multiprotein complex with the ERCC4(XPF)-ERCC1, MUS81-EME1, and SLX1 endonucleases, and also associates with MSH2/MSH3 mismatch repair complex, telomere binding complex TERF2(TRF2)-TERF2IP(RAP1), the protein kinase PLK1 and the uncharacterized protein C20orf94. The multiprotein complex is required for repair of specific types of DNA lesions and is critical for cellular responses to replication fork failure. The encoded protein acts as a docking platform for the assembly of multiple structure-specific endonucleases.
Specifications
| BTBD12 | |
| Polyclonal | |
| Unconjugated | |
| SLX4 | |
| BTB (POZ) domain containing 12; BTB/POZ domain-containing protein 12; BTBD12; FANCP; KIAA1784; KIAA1987; MUS312; SLX4; SLX4 structure-specific endonuclease subunit; SLX4 structure-specific endonuclease subunit homolog; structure-specific endonuclease subunit SLX4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84464 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IY92 | |
| SLX4 | |
| Synthetic peptide directed towards the N-terminal of human BTBD12 (aa 100-149). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction