Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTBD14A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982461
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2461 Supplier Bioss Supplier No. BS12907R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTBD5 is a transcription factor containing BTB (POZ) domain.

Specifications

Antigen BTBD14A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Klhl28
Gene Accession No. Q9NXS3
Gene Alias 2810440N09Rik; 4122402F11Rik; 4931401E10Rik; BOS_20502; BTB (POZ) domain containing 5; BTB (POZ) domain containing 5-like; BTB/POZ domain-containing protein 5; BTBD5; kelch like family member 28; kelch-like 28; kelch-like 28 (Drosophila); kelch-like family member 28; Kelch-like protein 28; KLHL28; LOC100341808
Gene Symbols Klhl28
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BTBD14A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54813
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.