Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTBD15 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178185B
Description
BTBD15 Polyclonal antibody specifically detects BTBD15 in Human samples. It is validated for Western BlotSpecifications
BTBD15 | |
Polyclonal | |
Western Blot | |
BTB (POZ) domain containing 15, BTB/POZ domain-containing protein 15, BTBD15, HSPC063, MGC57431, MGC60348, MGC88058, zinc finger and BTB domain containing 44, zinc finger and BTB domain-containing protein 44, Zinc finger protein 851, ZNF851MGC26123 | |
The immunogen for this product maps to a region between residue 350 and 400 of human Zinc Finger and BTB Domain Containing 44 using the numbering given in entry BAI46608.1 (GeneID 29068). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
29068 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction