Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTBD16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559208 Supplier Invitrogen™ Supplier No. PA559208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLDRDIGRSL RPLFLCLRLH GITKGKDLEV LRHLNFFPES WLDQVTVNHY HALENGGDMV HLKDLNTQAV RFGLLFNQEN TTYSKTIALY GFFFK Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

BTBD16 encodes a protein that contains a BTB/POZ domain. This domain mediates protein-protein interactions. A mutation in this gene may be associated with bipolar disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD16
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BTBD16
Gene Accession No. Q32M84
Gene Alias BTB (POZ) domain containing 16; BTB domain containing 16; BTB/POZ domain-containing protein 16; Btbd16; C10orf87; E330040A16Rik; FLJ25359; MGC129864; MGC129865
Gene Symbols BTBD16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BTBD16. Recombinant protein control fragment (Product #RP-97097).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118663
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.