Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTBD18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561933
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561933 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561933 Supplier Invitrogen™ Supplier No. PA561933
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPSEVSEVLS VGGRWTPDLE ITSSQPLDGQ EDKLLHVSSL DTPQRSYGDL SPPCSNWVET GLEVSLTTDE LLYPSPKAGK EVSGHSELLG SLPASS Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 68%.

Specifically required during spermatogenesis to promote expression of piRNA precursors. The piRNA metabolic process mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and governs the methylation and subsequent repression of transposons, which is essential for the germline integrity. Acts by facilitating transcription elongation at piRNA loci during pachytene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD18
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BTBD18
Gene Accession No. B2RXH4
Gene Alias BTB (POZ) domain containing 18; BTB domain containing 18; BTB/POZ domain-containing protein 18; BTBD18; hCG1730474
Gene Symbols BTBD18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BTBD18. Recombinant protein control fragment (Product #RP-100681).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643376
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.