Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTBD19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560060 Supplier Thermo Scientific Supplier No. PA560060
Add to Cart
Add to Cart

BTBD19 Polyclonal Antibody

BTBD19 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD19
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BTBD19
Gene Accession No. C9JJ37
Gene Alias 9530048O09Rik; BB115071; BTB (POZ) domain containing 19; BTB domain containing 19; BTB/POZ domain-containing protein 19; BTBD19; novel BTB domain containing protein
Gene Symbols BTBD19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BTBD19. Recombinant protein control fragment (Product #RP-99850).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149478
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.