Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTBD19 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988531
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8531 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8531 Supplier Bioss Supplier No. BS8401R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTBD19 is a protein coding gene.

Specifications

Antigen BTBD19
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BTBD19
Gene Accession No. C9JJ37
Gene Alias 9530048O09Rik; BB115071; BOS_3702; BTB (POZ) domain containing 19; BTB domain containing 19; BTB/POZ domain-containing protein 19; BTBD19; LOC100348658; novel BTB domain containing protein
Gene Symbols BTBD19
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BTBD19.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149478
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.