Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTBD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568639
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568639 Supplier Invitrogen™ Supplier No. PA568639
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 90%.

BTBD2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BTBD2
Gene Accession No. Q9BX70
Gene Alias 2610037C03Rik; 4930512K17Rik; BTB (POZ) domain containing 2; BTB domain containing 2; BTB/POZ domain-containing protein 2; BTBD2; LOW QUALITY PROTEIN: BTB/POZ domain-containing protein 2; RGD1566094
Gene Symbols BTBD2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human BTBD2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55643
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.