Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTBD3 Antibody (OTI1A10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271699H
Description
BTBD3 Monoclonal specifically detects BTBD3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
BTBD3 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
BTBD3 | |
Full length recombinant protein of human BTBD3 (NP_852108) produced in E.coli. | |
0.1 mL | |
22903.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1A10 | |
Western Blot | |
BTB (POZ) domain containing 3, BTB/POZ domain-containing protein 3, KIAA0952dJ742J24.1, MGC130038, MGC130039 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction