Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTBD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540965 Supplier Invitrogen™ Supplier No. PA540965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FLWYTAAKKP ELQFVSKARK GLVPQRCHRF QSCAYRSNQW RYRGRCDSIQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

BTBD3 is a protein containing BTB/POZ domain. Its function has not been determined yet.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BTBD3
Gene Accession No. Q9Y2F9
Gene Alias BTB (POZ) domain containing 3; BTB domain containing 3; BTB/POZ domain containing protein 3; BTB/POZ domain-containing protein 3; BTBD3; dJ742J24.1; KIAA0952; mKIAA0952
Gene Symbols BTBD3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human BTBD3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22903
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.