Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTBD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206279
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6279 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6279 Supplier Fabgennix Inc Supplier No. BTBD3301AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTBD3 Polyclonal antibody specifically detects BTBD3 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen BTBD3
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene BTBD3
Gene Accession No. P58545, Q9Y2F9
Gene Alias BTB (POZ) domain containing 3; BTB domain containing 3; BTB/POZ domain containing protein 3; BTB/POZ domain-containing protein 3; BTBD3; dJ742J24.1; KIAA0952; mKIAA0952
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 410-460 on human BTBD3 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228662, 22903, 311462
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.