Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTBD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541655 Supplier Invitrogen™ Supplier No. PA541655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FVVGPPGATR TVPAHKYVLA VGSSVFYAMF YGDLAEVKSE IHIPDVEPAA Sequence homology: Cow: 86%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Rabbit: 79%; Zebrafish: 79%.

Adapter protein for the CUL3 E3 ubiquitin-protein ligase complex. Involved in late neuronal development and muscle formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD6
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BTBD6
Gene Accession No. Q96KE9
Gene Alias BDPL; BTB (POZ) domain containing 6; BTB domain containing 6; BTB domain protein BDPL; BTB/POZ domain-containing protein 6; Btbd6; glucocorticoid receptor AF-1 coactivator-1; lens BTB domain protein; LOW QUALITY PROTEIN: BTB/POZ domain-containing protein 6
Gene Symbols BTBD6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human BTBD6 (aa 87-136).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90135
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.