Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTBD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556711
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556711 Supplier Thermo Scientific Supplier No. PA556711
Add to Cart
Add to Cart

BTBD6 Polyclonal Antibody

BTBD6 Polyclonal Antibody for Western Blot, ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BTBD6
Gene Accession No. Q96KE9
Gene Alias BDPL; BTB (POZ) domain containing 6; BTB domain containing 6; BTB domain protein BDPL; BTB/POZ domain-containing protein 6; Btbd6; glucocorticoid receptor AF-1 coactivator-1; lens BTB domain protein; LOW QUALITY PROTEIN: BTB/POZ domain-containing protein 6
Gene Symbols BTBD6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BTBD6. Recombinant protein control fragment (Product #RP-89144).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90135
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.