Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTBD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545934 Supplier Invitrogen™ Supplier No. PA545934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SPRVGGNSQA QQTFIGTSSY SQQGYGCESK LYSLDHGHEK PQDKKKRTSG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 82%; Zebrafish: 79%.

Acts as a mediator of epithelial dynamics and organ branching by promoting cleft progression. Induced following accumulation of fibronectin in forming clefts, leading to local expression of the cell-scattering SNAIL2 and suppression of E-cadherin levels, thereby altering cell morphology and reducing cell-cell adhesion. This stimulates cell separation at the base of forming clefts by local, dynamic intercellular gap formation and promotes cleft progression. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BTBD7
Gene Accession No. Q9P203
Gene Alias 5730507E09Rik; 8030448M07; BTB (POZ) domain containing 7; BTB domain containing 7; BTB/POZ domain-containing protein 7; Btbd7; E130118E17Rik; function-unkown protein 1; Fup1; Kiaa1525; mKIAA1525; si:dkeyp-55f12.1; wu:fd08g07
Gene Symbols BTBD7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human BTBD7 (aa 13-62).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55727
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.