Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTBD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532003
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532003 Supplier Invitrogen™ Supplier No. PA532003
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (98%), Rat (99%), Zebrafish (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Acts as a mediator of epithelial dynamics and organ branching by promoting cleft progression. Induced following accumulation of fibronectin in forming clefts, leading to local expression of the cell-scattering SNAIL2 and suppression of E-cadherin levels, thereby altering cell morphology and reducing cell-cell adhesion. This stimulates cell separation at the base of forming clefts by local, dynamic intercellular gap formation and promotes cleft progression. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene BTBD7
Gene Accession No. Q9P203
Gene Alias 5730507E09Rik; 8030448M07; BTB (POZ) domain containing 7; BTB domain containing 7; BTB/POZ domain-containing protein 7; Btbd7; E130118E17Rik; function-unkown protein 1; Fup1; Kiaa1525; mKIAA1525; si:dkeyp-55f12.1; wu:fd08g07
Gene Symbols BTBD7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BTBD7. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55727
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.