Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BTBD8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557220
Description
Immunogen sequence: LYVKPCCPDI DIFVDGKRFK AHRAILSARS SYFAAMLSGC WAESSQEYVT LQGISHVELN VMMHFIYGGT LDIPDKTNVG QILNMAD Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
BTBD8, also named as BTB/POZ domain-containing protein 8, is a 378 amino acid protein, which is highly expressed in fetal brain and localizes in the nucleus. BTBD8 exists two isoforms and the molecular weight of isoforms are 43 kDa and 35 kDa.
Specifications
BTBD8 | |
Polyclonal | |
Unconjugated | |
BTBD8 | |
BTB (POZ) domain containing 8; BTB domain containing 8; BTB/POZ domain-containing protein 8; BTBD8; double BTB/POZ domain containing protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284697 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5XKL5 | |
BTBD8 | |
Recombinant protein corresponding to Human BTBD8. Recombinant protein control fragment (Product #RP-94538). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction