Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTBD9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116850
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116850 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116850 Supplier Invitrogen™ Supplier No. PA5116850
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, U-251MG, SH-SY5Y Immunogen sequence: RDSRSYSYFI EVSMDELDWV RVIDHSQYLC RSWQKLYFPA RVCSGDGVSL WCPLWSRTPE LKQSSLLGLP KCRYIRIVGT HNTVNKIFHI VAFECMFTNK TFTLEKGLIV PMENVATIAD CASVIEGVSR SRNALLNGDT KNYDWDSGYT CHQLGSGAIV VQLAQPYMIG SIRLLLWDCD DRSYSYYVEV STNQQQWTMV ADRTKVSCKS WQSVTFERQP ASFIRIVGTH NTANEVFHCV HFECPEQQSS QKEENSEESG TGDTSLAGQQ LDSHALRAPS GSSLPSSPGS NSRSPNRQHQ.

This locus encodes a BTB/POZ domain-containing protein. This domain is known to be involved in protein-protein interactions. Polymorphisms at this locus have been reported to be associated with susceptibility to Restless Legs Syndrome and may also be associated with Tourette Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BTBD9
Gene Accession No. Q5PQR3, Q96Q07
Gene Alias 1700023F20Rik; 4930402L05; AI464421; BTB (POZ) domain containing 9; BTB domain containing 9; BTB/POZ domain-containing protein 9; Btbd9; dJ322I12.1; KIAA1880
Gene Symbols BTBD9
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114781, 294318
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.