Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTC, AM, cell permeant

Catalog No. 501147552
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-114-7552 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-114-7552 Supplier Invitrogen™ Supplier No. B6791
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

BTC, AM, cell permeant

BTC, AM is a cell-permeant coumarin benzothiazole-based Ca2+ indicator. It exhibits a shift in excitation maximum from about 480 nm to 400 nm upon binding Ca2+, which permits ratiometric measurements that are essentially independent of uneven dye loading cell thickness, photobleaching and dye leakage. BTC has high selectivity for Ca2+ and a Kd ∼7μM enabling quantitation of high intracellular Ca2+ levels.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Flow Cytometer
Product Type Calcium Indicator
Quantity 100 μg
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.