Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BTD
GREENER_CHOICESDP

Catalog No. 694050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-405-0 50 mg
69-401-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 69-405-0 Supplier Tocris Bioscience™ Supplier No. 6940/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective TRPC5 activator

BTD is a selective TRPC5 activator (EC50 = 1.4 μM in a calcium influx assay, 1.3 μM in whole cell patch clamp assay). Displays >14.5-fold selectivity for TRPC5 over other TRP channels (EC50 = 20.6 μM for TRPM8). Also activates TRPC1:5 and TRPC4:5 heteromers.

Specifications

Chemical Name or Material 6-Methyl-N-[3-(tricyclo[3.3.1.13,7]dec-1-yloxy)propyl]-4H-1,2,4-benzothiadiazine-3-propanamide-1,1-dioxide
CAS 896684-04-1
Quantity 50 mg
Molecular Formula C24H33N3O4S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.