Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562581
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562581 Supplier Invitrogen™ Supplier No. PA562581
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLDIYEQQVM TAAQKDVQII VFPEDGIHGF NFTRTSIYPF LDFMPSPQVV RWNPCLEPHR FNDTEVLQRL SCMAIRGDMF LVA Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.

Biotinidase functions to recycle biotin in the body by cleaving biocytin (biotin-epsilon-lysine), a normal product of carboxylase degradation, resulting in regeneration of free biotin. Biotinidase has also been shown to have biotinyl-transferase activity. Defects in the biotinidase gene cause multiple carboxylase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTD
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Btd
Gene Accession No. P43251
Gene Alias biotinase; Biotinidase; Btd; btd {ECO:0000312; EC 3.5.1.12; EMBL:AAH90017.1}
Gene Symbols Btd
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BTD. Recombinant protein control fragment (Product #RP-96439).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 686
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.