Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTF Antibody (M33-P5B11), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250118AF594
Description
BTF Monoclonal specifically detects BTF in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
BTF | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
BCLAF1 | |
Peptide Sequence - KYQGDGIVED (amino acids 900 - 909) | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
M33-P5B11 | |
Western Blot, ELISA, Immunohistochemistry | |
bcl-2-associated transcription factor 1, BCL2-associated transcription factor 1, Btf, BTFKIAA0164bK211L9.1 | |
Mouse | |
Protein A purified | |
Apoptosis, DNA Repair | |
9774.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction