Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTF Antibody (M33-P5B11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | BTF |
---|---|
Clone | M33-P5B11 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Description
BTF Monoclonal antibody specifically detects BTF in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
BTF | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Apoptosis, DNA Repair | |
PBS | |
9774 | |
IgG1 κ | |
Protein A purified |
M33-P5B11 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
bcl-2-associated transcription factor 1, BCL2-associated transcription factor 1, Btf, BTFKIAA0164bK211L9.1 | |
Peptide Sequence - KYQGDGIVED (amino acids 900 - 909) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title