Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTF Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen BTF
Concentration 0.5 mg/mL
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224859SS
SDP
View Documents
Novus Biologicals
NBP224859SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP224859
SDP
View Documents
Novus Biologicals
NBP224859
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

BTF Polyclonal specifically detects BTF in Human, Mouse, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

BTF
Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Polyclonal
Rabbit
Apoptosis, DNA Repair
0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
bcl-2-associated transcription factor 1, BCL2-associated transcription factor 1, Btf, BTFKIAA0164bK211L9.1
BCLAF1
IgG
Affinity Purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in human and chimpanzee, and 87% homologous in orangutan.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Canine, Primate
Q9NYF8
9774
A portion of amino acids 700-750 of human BTF was used as the immunogen for this antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.