Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527424 Supplier Invitrogen™ Supplier No. PA527424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, H1299, HeLaS3, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (96%), Xenopus laevis (97%), Dog (100%), Pig (100%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the basic transcription factor 3. This protein forms a stable complex with RNA polymerase IIB and is required for transcriptional initiation. Alternative splicing results in multiple transcript variants encoding different isoforms. This gene has multiple pseudogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTF3
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene BTF3
Gene Accession No. P20290
Gene Alias 1700054E11Rik; basic transcription factor 3; BETA-NAC; BTF3; BTF3a; BTF3b; NACB; NAC-beta; Nascent polypeptide-associated complex subunit beta; nascent-polypeptide-associated complex beta polypeptide; OK/SW-cl.8; RNA polymerase B transcription factor 3; transcription factor BTF3
Gene Symbols BTF3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BTF3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 689
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.