Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTF3L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568700 Supplier Invitrogen™ Supplier No. PA568700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTF3L1 Polyclonal Antibody for Western Blot

BTF3P11 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTF3L1
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BTF3P11
Gene Accession No. 0
Gene Alias basic transcription factor 3 pseudogene 11; BTF3L1; BTF3P11; HUMBTFB; OCIF; OPG; TNFRSF11B
Gene Symbols BTF3P11
Host Species Rabbit
Immunogen synthetic peptide directed towards the n terminal region of human BTF3L1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 690
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.