Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTF3L4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104189 Supplier Invitrogen™ Supplier No. PA5104189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BTF3L4.

The initiation of gene transcription involves the ordered assembly of a multiprotein complex on proximal promoter elements such as the TATA box. In addition to RNA polymerase II (Pol II), the transcription factor class II (TFII) family of proteins are required for initiation of transcription, as the first step in the formation of this initiation complex is the stable binding of TFIID to the TATA box. An additional TFII-related protein, BTF3, does not directly associate with the proximal promoter, but rather forms a stable complex with Pol II and facilitates Pol II assembly into the complex. The BTF3 gene is ubiquitously expressed and encodes two protein isoforms, BTF3a and BTF3b, which are produced from alternative splicing. The BTF3 proteins are identical, except that BTF3b lacks the first 44 amino acids at the N-terminal of BTF3a. As a consequence of this deletion, BTF3b is unable to induce transcription, despite being able to bind Pol II. Additionally, BTF3a and BTF3b associate with the widely expressed protein kinase casein kinase II. Casein kinase II phosphorylates BTF3a, as well as TFIIB, and is required for the efficient transcription of the tRNA and 55 rRNA genes by Pol III. BTF3 belongs to the NAC-beta family, which includes several related proteins, such as BTF3L1, BTF3L2, BTF3L3 and BTF3L4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTF3L4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Btf3l4
Gene Accession No. Q96K17, Q9CQH7
Gene Alias 4632412E09Rik; 5730434I03Rik; AI316863; AU022207; AW552446; basic transcription factor 3 like 4; basic transcription factor 3-like 4; BTF3L4; RGD1311774; Transcription factor BTF3 homolog 4
Gene Symbols Btf3l4
Host Species Rabbit
Immunogen A synthesized peptide derived from human BTF3L4(Accession Q96K17), corresponding to amino acid residues E96-V146.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366434, 70533, 91408
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.